Learn More
Medchemexpress LLC Apcin-A | 1683617-62-0 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13084110MM/1ML
Encompass_Preferred
Apcin-A is a small molecule inhibitor that selectively targets the cell division cycle protein Cdc20, a derivative of Apcin. It competitively binds to the D-box binding pocket of Cdc20, inhibiting substrate ubiquitination mediated by the anaphase promoting complex APC/C-Cdc20. This action prevents Cdc20 from binding to substrates like securin and cyclin B1, hindering anaphase initiation and cell cycle exit.
  • Selectively targets cell division cycle protein Cdc20.
  • Inhibits substrate ubiquitination mediated by APC/C-Cdc20.
  • Prevents Cdc20 binding to securin and cyclin B1.
  • Hindrance of anaphase initiation and cell cycle exit.
  • Potential for anti-mitotic drug development.
  • Potential to overcome tumor chemotherapy resistance.
  • Can be used in PROTAC synthesis.
Catalog No. 50-003-40655