Learn More
Medchemexpress LLC Apcin-A | 1683617-62-0 | 98.0% | 342.61 g/mol | C10H14Cl3N5O2 | 5 MG

Supplier: Medchemexpress LLC HY1308415MG
Apcin-A is a small-molecule inhibitor that selectively targets the APC/C co-activator Cdc20 and is used in biochemical and cell-based research to study cell cycle regulation and ubiquitin-mediated proteolysis. The compound is supplied as a high-purity solid with recommended cold-storage conditions for stability.
- Selective inhibitor of Cdc20-mediated APC/C activity.
- Purity 98.0%.
- Molecular weight 342.61 g/mol; formula C10H14Cl3N5O2.
- Available in multiple research sizes, including 5 mg.
- Suitable for biochemical assays and cell-based studies.
- Store powder at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.