Learn More
Enzo Life Sciences Apurinic / apyrimidinic endonuclease (human) monoclonal antibody (13B 8E5C2) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804213R100
Encompass_Preferred
Clone: 13B 8E5C2. Isotype: IgG2a. Immunogen: Recombinant human APE1 (apurinic/apyrimidinic endonuclease). Species reactivity: Human. Applications: IHC (PS), IP, WB. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9830