Learn More
Selleck Chemical LLC Apelin-13 P1211-25MG
SDP
Supplier:  Selleck Chemical LLC P121125MG
Encompass_Preferred
Apelin-13 can efficiently activate the APJ receptor with EC50 of 0 37 nM and result in suppress the forskolin-stimulated cAMP production
Catalog No. 50-000-40910