Learn More
Cayman Chemical ApelIn-13trIfluoroACTAte25mg
SDP
Supplier:  Cayman Chemical 1352325 MG
Encompass_Preferred
Endogenous ligand of the APJ receptor, with an EC50 of 0.37 nM; acts primarily in the periphery and CNS, playing important roles in regulating cardiovascular function, fluid homeostasis, hypertension, and insulin sensitivity
Catalog No. 50-295-5657