Learn More
Selleck Chemical LLC APG-2575 S9970-25MG
SDP
Supplier:  Selleck Chemical LLC S997025MG
Encompass_Preferred
APG-2575 (lisaftoclax) is a dual Bcl-2 and Bcl-xl inhibitor with IC50 values of 2 nM and 5 9 nM for Bcl-2 and Bcl-xl respectively
Catalog No. 50-000-24440