Learn More
Enzo Life Sciences (+)-Aphidicolin (1mg). CAS: 38966-21-1
SDP
Supplier:  Enzo Life Sciences BMLCC1010001
Encompass_Preferred
Produced by Cephalosporium aphidicola and other fungi. A diterpene fungal metabolite that acts as reversible inhibitor of eukaryotic nuclear DNA replication. Useful for cell synchronization. Blocks the cell cycle at early S phase. Prolongs the half life of DNA methyltransferase. Specific inhibitor of DNA polymerase α and δ in eukaryotic cells and in some viruses of animal origin. Acts synergistically with vincristine and doxorubicin. Apoptosis inhibitor/inducer. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0402