Learn More
Abcam Aphidicolin, DNA polymerase alpha, delta and epsilon inhibitor, 1MG
Supplier:  Abcam AB1424001MG
Encompass_Preferred
MW 338.5 Da, Purity >98%. DNA polymerase α, δ and ε inhibitor. Tetracyclic diterpene antibiotic. Reversible inhibitor of eukaryotic nuclear DNA replication. It blocks the cell cycle at early S phase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3319