Learn More
Apexbio Technology LLC Apicidin 183506-66-3 1mg
SDP
Supplier:  Apexbio Technology LLC A81761
Encompass_Preferred
Apicidin (CAS 183506-66-3) is a small-molecule inhibitor targeting histone deacetylases (HDACs) It is designed to inhibit HDAC activity thereby modulating gene expression through altered chromatin structure Apicidin exerts its biological activity primarily through selective inhibition of HDAC enzymatic function In cell-based and in vitro studies Apicidin demonstrates antiproliferative and antiangiogenic activity with an IC50 of approximately 0 7 1 1 nM in parasitic organisms Based on these pharmacological properties Apicidin holds research potential in oncology and parasitology
Catalog No. 50-000-92114