Learn More
Enzo Life Sciences Apigenin (100mg). CAS: 520-36-5
SDP
Supplier:  Enzo Life Sciences BMLEI3450100
Encompass_Preferred
Apigenin is an antioxidant flavonoid. Has chemopreventive and antitumor  properties. Induces apoptosis. Inhibits the proliferation of malignant tumor cells by G2/M arrest. Is a MAP-Kinase (MAPK/ERK) inhibitor. Inhibits hypoxia-inducible factor-1 (HIF-1) and vascular endothelial growth factor (VEGF) expression. Induces gastric relaxation in mouse stomach, mainly by suppressing Ca2+ influx through voltage-dependent calcium channels. Apigenin is a selective inhibitor of CK2. In WEHI 231 cells inhibition of CK2 by apigenin decreased the rate of IκBα turnover and nuclear levels of NF-κB (20 µM). It blocks proliferation of Wnt-1 transfected cells and abrogates phosphorylation of β-catenin. In a rat model of glomerulonephritis, inhibition of CK2 by apigenin ameliorates renal dysfunction. Purity: ≥97% (HPLC). Solubility: Soluble in  diluted potassium hydroxyde or DMSO, slightly soluble in 100% ethanol (hot). Insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0732