Learn More
Apexbio Technology LLC Apigenin, 10mM (in 1mL DMSO) CAS# 520-36-5
SDP
Supplier:  Apexbio Technology LLC N18285.1
Encompass_Preferred
Apigenin is an antioxidant plant flavanoid which contains anti-inflammatory and anticancer properties. It may induce apoptosis and may inhibit proliferation of tumor cells by arresting the cell cycle at the G2/M phase. Is also known to be a MAP kinase inh Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8344
 Restricted Item