Learn More
Cayman Chemical ApIgenIn 7-glucosIde 5mg
SDP
Supplier:  Cayman Chemical 268135 MG
Encompass_Preferred
A flavonoid with anti-inflammatory and anxiolytic activities; inhibits LPS-induced nitric oxide production in RAW 264.7 cells from 0.16-10 μM; reduces pulmonary edema and lung inflammation in a mouse model of LPS-induced acute lung injury at 10 mg/kg; increases the number of entries and the time spent in the open arms of the elevated plus maze in rats
Catalog No. 50-293-2692