Learn More
Cayman Chemical Apigenin Inhibitors
SDP
Supplier:  Cayman Chemical 1001027525 MG
Encompass_Preferred
Inhibits CK2 activity in the renal cortex with an IC50 value of 30 µM; potent inhibitor of NO and PGE2 biosynthesis by reducing iNOS and COX-2 expression
Catalog No. 50-288-9522