Learn More
Medchemexpress LLC Apilimod | 541550-19-0 | 99.8% | 418.49 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14644100MG
Encompass_Preferred
Apilimod (STA 5326) is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC50s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. It is a potent and highly selective PIKfyve inhibitor.
  • Potent IL-12/IL-23 inhibitor
  • Highly selective PIKfyve inhibitor
  • Inhibits IL-12 with low IC50 values
  • Used in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs
Catalog No. 50-003-60121