Learn More
Medchemexpress LLC HY-14644A 100mg , Apilimod (mesylate) CAS:870087-36-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14644A100MG
Encompass_Preferred
HY-14644A 100mg Apilimod (mesylate) CAS: 870087-36-8 Apilimod (STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC50s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. Apilimod is a potent and highly selective PIKfyve inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0135