Learn More
Medchemexpress LLC Apilimod (mesylate) | 870087-36-8 | 99.8% | 610.70 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14644A10MM/1ML
Encompass_Preferred
Apilimod (STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, strongly inhibiting IL-12 with IC50s of 1 nM and 2 nM in IFN-γ/SAC-stimulated human and monkey PBMCs. It is also a potent and highly selective PIKfyve inhibitor.
  • Potent IL-12/IL-23 inhibitor
  • Strongly inhibits IL-12
  • Potent and highly selective PIKfyve inhibitor
  • Inhibits IFN-γ production in human PBMCs
  • Reduces severity of experimental autoimmune uveoretinitis (EAU)
Catalog No. 50-003-63816