Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apiopaeonoside | 100291-86-9 | MFCD29049468 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYN216110MM/1ML

Encompass_Preferred

Apiopaeonoside is a natural product isolated from the root of Paeonia suffruticosa. This white to off-white solid has a molecular weight of 460.43 and a chemical formula of C20H28O12, with a purity of 99.81%. It is classified under phenols and is intended for research use only.

  • Isolated from the root of Paeonia suffruticosa
  • White to off-white solid
  • Molecular weight of 460.43
  • Chemical formula: C20H28O12
  • Purity of 99.81%
  • Soluble in DMSO at 100 mg/mL
  • Storage as solid: 4°C, sealed, away from moisture and light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture and light

Catalog No. 50-004-21729


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.