Learn More
Medchemexpress LLC APLNApelin Mouse 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7756350UG
Encompass_Preferred
APLNApelin protein is the endogenous ligand of APLNR and drives APLNR internalization APLNApelin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived APLNApelin protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-87047