Learn More
Medchemexpress LLC Apn-c3-peg4-azide | 2183440-32-4 | 97.97% | C24H32N6O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140841100MG
Encompass_Preferred
APN-C3-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Used in PROTAC synthesis for selective degradation of target proteins.
Catalog No. 50-003-55567