Learn More
Medchemexpress LLC APN-C3-PEG4-azide | 2183440-32-4 | 97.97% | C24H32N6O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14084150MG
Encompass_Preferred
APN-C3-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-53136