Learn More
Medchemexpress LLC APN-PEG5-VC-PAB-MMAE | 2252392-31-5 | 95.19% | 1577.90 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1635495MG
Encompass_Preferred
APN-PEG5-VC-PAB-MMAE is a drug-linker conjugate that is part of INA03, a transferrin-competitive antibody-drug conjugate against CD71. It is for research use only.
  • Drug-linker conjugate
  • Part of INA03 (a transferrin-competitive antibody-drug conjugate against CD71)
Catalog No. 50-003-92970