Learn More
Medchemexpress LLC Apn-peg5-vc-pab-mmae | 2252392-31-5 | 95.2% | 1577.90 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1635491MG
Encompass_Preferred
APN-PEG5-VC-PAB-MMAE is a drug-linker conjugate that is part of INA03, a transferrin-competitive antibody-drug conjugate against CD71. Antibody-drug conjugates (ADCs) are comprised of an antibody to which an ADC cytotoxin is attached through an ADC linker. This product is for research use only.
  • Drug-linker conjugate
  • Component of INA03, a transferrin-competitive antibody-drug conjugate against CD71
  • Used in antibody-drug conjugates (ADCs)
Catalog No. 50-003-92526