Learn More
Medchemexpress LLC β-Apo-13-carotenone (D'Orenone) | 17974-57-1 | MFCD31544340 | 96.5% | 258.40 | C18H26O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10195310MG
Encompass_Preferred
β-Apo-13-carotenone (D'Orenone) is a naturally occurring β-apocarotenoid used in research as an antagonist of the retinoid X receptor alpha (RXRα). It is supplied as a light yellow oil with documented DMSO solubility and is intended for laboratory research use only.
  • Acts as an antagonist of RXRα.
  • Naturally occurring β-apocarotenoid.
  • Light yellow oil appearance.
  • High purity (~96.5%) as reported in product documentation.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic recommended.
  • Requires storage at -80°C and protection from light.
  • Supplied in small quantities appropriate for in vitro research.
Catalog No. 50-002-47242