Learn More
ACROBiosystems Recombinant Protein;Human Apo-A1 Protein;HEK293;50UG;AP1,Apolipoprotein A-I,Apo-AI,ApoA-I,APOA1,MGC117399,APO-A1,Apolipoprotein A-1,Apolipoprotein A1,ApoA1
Supplier:  ACROBiosystems AP1H522550UG
Encompass_Preferred
Recombinant Protein;Human Apo-A1 Protein;50UG;Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It contains AA Arg 19 - Gln 267 (Accession # NP_000030.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.8 kDa. The protein migrates as 26-30 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9049