Learn More
New England Biolabs, Inc. APOBEC3A - 4000 units
SDP
Supplier:  New England Biolabs, Inc. M0648L
Encompass
APOBEC3A (Apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A) is a cytidine deaminase enzyme that catalyzes the deamination of cytosine or 5-methylcytosine in single-stranded DNA to produce uracil or thymine.
  • Ideal for cytosine deaminationIdeal for APOBEC-Coupled Epigenetic Sequencing (ACE-Seq)
  • Isolated from a recombinant source
  • Supplied with 10X Reaction Buffer
Catalog No. 50-003-32610