Learn More
Medchemexpress LLC Apolipoprotein A-I (APOA1) | 10 UG

Supplier: Medchemexpress LLC HYP7550310UG
Recombinant mouse apolipoprotein A-I, His-tagged, expressed in HEK293 cells and supplied as a 0.22 μm filtered solution for research into lipid metabolism and cholesterol transport. The product is formulated in PBS (pH 7.4) with stabilizers and is provided with low endotoxin and recommended frozen storage for long-term stability.
- Recombinant His-tagged mouse apolipoprotein A-I expressed in HEK293 cells.
- Supplied as a 0.22 μm filtered solution in PBS, pH 7.4.
- Formulated with 5% trehalose, 5% mannitol, and 0.01% Tween 80 for stability.
- Molecular weight approximately 27-31 kDa.
- Endotoxin level <1 EU/μg by LAL method.
- Store at -80°C and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.