Learn More
Medchemexpress LLC Apolipoprotein A-I (APOA1) | 97.0% | 5 UG

Supplier: Medchemexpress LLC HYP75503A5UG
Recombinant mouse apolipoprotein A-I expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified protein for research use, it is suitable for biochemical assays, functional studies of HDL/cholesterol efflux, and use as an assay control.
- Recombinant mouse apolipoprotein A-I expressed in HEK293 cells
- C-terminal His tag for detection and purification
- High purity (≈97.0%) suitable for biochemical assays
- Molecular weight approximately 26-28 kDa
- Suitable for in vitro functional studies and assay controls
- Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.