Learn More
Medchemexpress LLC Apolipoprotein A-I (mouse) | 97.0% | 100 UG

Supplier: Medchemexpress LLC HYP75503A100UG
Recombinant mouse apolipoprotein A-I produced in HEK293 cells, supplied as a lyophilized protein with a C-terminal 6xHis tag. Intended for biochemical and immunological assays related to HDL function and cholesterol transport. Lyophilized from PBS (pH 7.4) with analytical data provided in a certificate of analysis.
- Recombinant protein produced in HEK293 cells.
- C-terminal 6xHis tag for purification and detection.
- Purity 97.0% as stated in certificate of analysis.
- Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4.
- Not recommended to reconstitute below 100 μg/mL.
- Store at -20°C for long-term stability; freeze aliquots at -20°C or -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.