Learn More
Medchemexpress LLC Apolipoprotein E/APOE protein, rat (His) | >90.0% | 100UG

Supplier: Medchemexpress LLC HYP701096100UG
Recombinant rat apolipoprotein E (APOE) is supplied as an N-6xHis-tagged protein expressed in E. coli and provided lyophilized. It is involved in lipid transport and lipoprotein metabolism; the product covers residues E19-Q312, has an approximate molecular weight of 45 kDa, and is provided with documentation for handling and quality.
- Recombinant rat APOE spanning E19-Q312 with N-6xHis tag.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Lyophilized formulation stabilized with trehalose for storage.
- Available in multiple sizes to suit experimental needs.
- Includes datasheet, certificate of analysis, and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.