Learn More
Medchemexpress LLC Apolipoprotein E (APOE2; R154S, R176C), human, C-terminal 6xHis | >90.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP70109410UG
Encompass_Preferred
Recombinant human apolipoprotein E (APOE2 variant, R154S, R176C) expressed in HEK293 cells and supplied with a C-terminal 6xHis tag. The protein is glycosylated and purified for use in biochemical assays related to lipid metabolism and neurodegeneration research; formulation concentration is not specified on the product page.
Catalog No. 50-001-76357