Learn More
Medchemexpress LLC Apolipoprotein E/APO 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7854250UG
Encompass_Preferred
Apolipoprotein E (APOE) plays a key role in lipoprotein-mediated lipid transport and is a core component in the production transformation and clearance of plasma lipoproteins As an amphipathic molecule APOE binds to various lipoprotein particles including chylomicrons chylomicron remnants VLDL and IDL favoring HDL Apolipoprotein E/APOE Protein Human (C130R HEK293 Fc) is the recombinant human-derived Apolipoprotein E/APOE protein expressed by HEK293 with N-hFc labeled tag and C130R mutation
Catalog No. 50-001-94064