Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein E (APOE2) protein, human (R154S, R176C) | >90.0% | 5UG
SDP

Supplier:  Medchemexpress LLC HYP7010945UG

Encompass_Preferred

Recombinant human apolipoprotein E (APOE2 variant R154S, R176C) provided as a purified protein produced in HEK293 cells with a C-terminal His tag. The material is intended for in vitro biochemical characterization, binding studies, and antibody validation where isoform-specific APOE2 activity is required.

  • Produced in HEK293 expression system.
  • Contains APOE2-specific R154S and R176C substitutions.
  • C-terminal His tag for detection and purification.
  • Purity >90% as determined by reducing SDS-PAGE.
  • Available in small research pack sizes (for example, 5 μg).
  • Suitable for biochemical assays and antibody validation.

Catalog No. 50-001-75923


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.