Learn More
Medchemexpress LLC Apolipoprotein E/APOE3 protein, human (HEK293, His) | >95.0% | 5 UG
SDP
Supplier:  Medchemexpress LLC HYP75315UG
Encompass_Preferred
Recombinant human apolipoprotein E (APOE3) produced in HEK293 cells with an N-terminal His tag for research use. The protein is glycosylated (apparent MW ~36-40 kDa) and supplied in small laboratory quantities. Follow storage and reconstitution recommendations to preserve integrity.
Catalog No. 50-002-92566