Learn More
Medchemexpress LLC HY-P7535 10ug Medchemexpress, Apolipoprotein J, C-Fc-His (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP753510UG
Encompass_Preferred
Medchemexpress, HY-P7535 10ug Apolipoprotein J, C-Fc-His (HEK293-expressed), Human CAS: Recombinant Human Apolipoprotein J, C-Fc-His (HEK293-expressed)expresses in HEK293 with an Fc fragment and a His tag at the N-terminus. Apolipoprotein J (ApoJ) is involved in numerous physiological process important for lipid transportation, vascular smooth muscle cell differentiation, and has the potential for atherosclerosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3176