Learn More
Cayman Chemical ApoptolIdIn 100ug
SDP
Supplier:  Cayman Chemical 19435100 UG
Encompass_Preferred
An apoptosis inducer that inhibits the mitochondrial F1FO ATPase (Ki = 4-5 µM); selectively kills transformed glial cells without significantly affecting untransformed cells
Catalog No. 50-298-2366