Learn More
Biotium Apoptosis Assays Fluorescent Proteins Nucleotides Other Conjugates Annexin V Conjugates CF740 1/EA
Supplier:  Biotium 29126
Encompass_Preferred
Annexin V fluorescent conjugates can be used to detect apoptotic cells The human anticoagulant Annexin V is a 35-36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS) This product is labeled with near-infrared CF740 Dye (Ex/Em 742/762 nm)
Catalog No. 50-005-00240