Learn More
Enzo Life Sciences Apoptosis inducer set (1 Set)
SDP
Supplier:  Enzo Life Sciences ALX850235KI01
Encompass_Preferred
The set contains 5 ready-to-use reagents that induce apoptosis through different mechanisms. Actinomycin D, an antineoplastic antibiotic, inhibits RNA synthesis. Camptothecin, an inhibitor of nuclear topoisomerase, induces apoptosis in many types of cells. Cycloheximide, an active antibiotic against many yeast and fungi, inhibits protein synthesis. Dexamethasone, an active and highly stable glucocorticoid, probably induces apoptosis by binding and activating the intracellular glucocorticoid receptor. Etoposide, a derivative of podophyllotoxin, inhibits topoisomerase activity. Long Term Storage: -20°C.
Catalog No. 50-201-0124