Learn More
Biotium Apoptotic, Necrotic & Healthy Cells Quantification Kit (50 assays)
Supplier:  Biotium 30018
Encompass_Preferred
This kit provides a convenient assay for quantifying apoptotic (green) and necrotic (red) cells within the same cell population by flow cytometry or fluorescence microscopy. The kit contains FITC-Annexin V which binds to apoptotic cells by binding to phosphatidylserine (PS) exposed on the cell surface, and Ethidium Homodimer III (EthD-III) which is a membrane impermeant nucleic acid probe specific for dead cells. Staining is rapid, requiring only a single 15-30 minute staining step. FITC-Annexin V has an excitation emission of 498/517 nm, and EthD-III has an excitation/emission of 522/593 nm (and a large absorption peak at 279 nm) when bound to DNA. The kit also contains a membrane permeable DNA dye Hocechst 3342 to stain all cell nuclei blue for analysis of healthy cells. The kit has reagents sufficient reagent for 50 assays.
Catalog No. 50-196-4474