Learn More
Medchemexpress LLC HY-15098 5mg Medchemexpress, Apoptozole CAS:1054543-47-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY150985MG
Encompass_Preferred
Medchemexpress, HY-15098 5mg Apoptozole CAS:1054543-47-3 Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0581