Learn More
Medchemexpress LLC Apostatin-1 | 2559703-06-7 | 99.8% | 345.50 | C19H27N3OS | 5 MG

Supplier: Medchemexpress LLC HY1340505MG
Apostatin-1 is a small-molecule research compound that potently inhibits TRADD-mediated signaling, modulates ubiquitination pathways, induces autophagy, and blocks apoptosis in cellular and animal models. It is supplied as a white to off-white solid with manufacturer-recommended storage and solubility guidance for laboratory use.
- Potent TRADD inhibitor that disrupts TRADD interactions.
- Binds TRADD-N with KD = 2.17 μM.
- Inhibits bortezomib-induced apoptosis (IC50 = 0.97 μM).
- Induces autophagy and promotes degradation of long-lived proteins at 10 μM (6 h).
- Soluble in DMSO at 83.33 mg/mL with ultrasonic warming and pH adjustment as recommended.
- Stable as a powder at -20°C for up to 3 years; store solutions frozen per guidance.
- High purity (>99.8%) suitable for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.