Learn More
Cayman Chemical APPC99 FragmentC-Term P 500uL
SDP
Supplier:  Cayman Chemical 28637500 UL
Encompass_Preferred
An analytical reference standard categorized as an intermediate in the phytocannabinoid biosynthetic pathway a precursor in the synthesis of CBDV CBGV and CBV intended for research and forensic applications
Catalog No. 50-293-9315