Learn More
Med Vet International Duraprep Solution Applicator 6ML
Supplier:  Med Vet International MMM8635H
Encompass_Preferred
Iodine povacrylex, one of the active ingredients in DuraPrep solution, has special attributes not found in povidone iodine. DuraPrep solution is persistent at least 48 hours against resident bacteria after a blood and saline (fluid) challenge. Surgical teams should apply surgical preps accordingly to manufacturer's directions. Each individual wrapped sterile applicator preps at least an 8" x 10" area. Each.
Catalog No. 50-304-5154