Learn More
Selleck Chemical LLC APR-246 (PRIMA-1MET) 5mg 5291-32-7
SDP
Supplier:  Selleck Chemical LLC S77245MG
Encompass_Preferred
APR-246, also known as PRIMA-1MET, is a small organic molecule that has been shown to restore tumour-suppressor function primarily to mutant p53 and also to induce cell death in various cancer types. APR-246 induces apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5179