Learn More
Medchemexpress LLC HY-119307 1mg Medchemexpress, Apratastat CAS:287405-51-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1193071MG
Encompass_Preferred
Medchemexpress, HY-119307 1mg Apratastat CAS:287405-51-0 Apratastat is an orally active, potent, and reversible dual inhibitor of tumor necrosis factor-α converting enzyme (TACE) and matrix metalloproteinases (MMPs) . Apratastat can potently inhibit the release of TNF-α in vitro, ex vivo, and in vivo with IC 50 s of 144 ng/mL in vitro and 81.7 ng/mL ex vivo, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2536