Learn More
Selleck Chemical LLC Aprepitant S1189-100mg
SDP
Supplier:  Selleck Chemical LLC S1189100MG
Encompass_Preferred
Aprepitant is a potent and selective neurokinin-1 receptor antagonist with IC50 of 0 1 nM Aprepitant reduces levels of pro-inflammatory cytokines including G-CSF IL-6 IL-8 and TNF Aprepitant inhibits HIV infection of human macrophages
Catalog No. 50-310-8261