Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aprepitant impurity 1 | 170729-76-7 | 95.75% | 50 MG
SDP

Supplier:  Medchemexpress LLC HY13077550MG

Encompass_Preferred

Aprepitant impurity 1 is an impurity of Aprepitant.

  • Molecular formula: C23H22F6N4O3
  • Molecular weight: 516.44
  • Target: Drug intermediate
  • Pathway: Others
  • Appearance: White to off-white solid
  • Purity: 95.75% by HPLC
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-40615


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.