Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aprepitant impurity 1 | 170729-76-7 | MFCD24386573 | 95.8% | 516.44 | C23H22F6N4O3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1307755MG

Encompass_Preferred

Aprepitant impurity 1 is an analytical impurity standard related to the NK-1 receptor antagonist aprepitant, supplied for impurity profiling, method validation, and analytical research. The compound is provided as a purified chemical standard with batch-specific documentation; consult the certificate of analysis and safety data sheet for storage, purity, and handling details.

  • Intended for impurity profiling and method validation.
  • Analytical standard suitable for HPLC and other chromatographic assays.
  • Molecular formula C23H22F6N4O3; molecular weight 516.44.
  • Purity 95.8% by HPLC (batch-specific values available in the COA).
  • Provided with batch certificate of analysis and safety data sheet.
  • For research use only; not for human consumption or clinical use.

Catalog No. 50-002-62918


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.