Learn More
Enzo Life Sciences APRIL (human) monoclonal antibody (Aprily-2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804844C100
Encompass_Preferred
Clone: Aprily-2. Isotype: IgG1. Immunogen: Recombinant human APRIL (aa 93-233). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: IHC (PS). Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9998