Learn More
Essendant San Jamar Bestguard Oven Mitt, 17", WebGuard Plus Fabric, Tan, Pair
Supplier:  Essendant SJM800FG17
Encompass_Preferred
Bestguard Ovenmitt with reinforced WebGuard Plus fabric between thumb and forefinger offers heat protection up to 450 F for 30 seconds Durable Kevlar stitching ensures insulation stays in place for maximum protection against flame heat and steam Size Group- Universal Material(s)- WebGuard Plus Fabric Color(s)- Tan Wrist Style- Angled Cuff
Catalog No. 50-000-26721