Learn More
Essendant APRON28X40KLENGRDWH
Supplier:  Essendant ANS36550
Encompass_Preferred
Universal size apron made of a breathable barrier SMS fabric which helps protect against hazardous and non-hazardous dust Strong and abrasion-resistant for extended wear Apparel Type- Apron Material(s)- MICROFORCE Barrier SMS Fabric Color(s)- White Color Family- White
Catalog No. 50-000-20413